siRNA Details
Gene Symbol: FMNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175736.4 | NP_783863.4 | 21 | 2716 | ||
NM_198900.2 | NP_944489.2 | 20 | 2563 | |||
XM_005269218.2 | XP_005269275.1 | 21 | 2695 | |||
XM_011538968.2 | XP_011537270.1 | 21 | 3185 | |||
XM_011538969.2 | XP_011537271.1 | 21 | 3184 | |||
XM_011538970.2 | XP_011537272.1 | 21 | 3184 | |||
XM_011538971.2 | XP_011537273.1 | 21 | 2760 | |||
XM_011538972.2 | XP_011537274.1 | 20 | 3030 | |||
XM_011538973.2 | XP_011537275.2 | 17 | 2175 | |||
XM_011538974.2 | XP_011537276.1 | 22 | 2517 | |||
GenBank mRNA | AB095934.2 | - | 21 | 2716 | ||
AK127200.1 | - | 8 | 1189 | |||
AK128195.1 | - | 20 | 2543 | |||
AL162062.1 | - | 5 | 673 | |||
BC033181.1 | - | 5 | 500 | |||
BC159100.1 | - | 21 | 2716 | 8860524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
