siRNA Details
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014788.3 | NP_055603.2 | 2 | 437 | ||
NM_033219.2 | NP_150088.1 | 2 | 437 | |||
XM_005252320.3 | XP_005252377.1 | 2 | 502 | |||
XM_006717338.1 | XP_006717401.1 | 2 | 1060 | |||
XM_011519244.2 | XP_011517546.1 | 2 | 267 | |||
XM_017015352.1 | XP_016870841.1 | 2 | 502 | |||
XM_017015353.1 | XP_016870842.1 | 2 | 502 | |||
XM_017015354.1 | XP_016870843.1 | 2 | 493 | |||
GenBank mRNA | AF220130.1 | - | 2 | 282 | ||
AF220131.1 | - | 2 | 282 | |||
AK055833.1 | - | 2 | 310 | |||
AK292825.1 | - | 2 | 304 | |||
BC006333.2 | - | 2 | 290 | 4299815 | ||
BF797334.1 | - | 2 | 395 | |||
D50919.1 | - | 2 | 282 | |||
DC405577.1 | - | 2 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
