siRNA Details
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 13 | 1547 | ||
NM_207325.2 | NP_997208.2 | 13 | 1573 | |||
XM_006723045.1 | XP_006723108.1 | 10 | 1089 | |||
XM_011526526.2 | XP_011524828.1 | 13 | 1528 | |||
XM_017026366.1 | XP_016881855.1 | 12 | 1462 | |||
XM_017026367.1 | XP_016881856.1 | 13 | 1487 | |||
GenBank mRNA | AK126264.1 | - | 7 | 648 | ||
AK126757.1 | - | 13 | 1564 | |||
AK129497.1 | - | 13 | 1528 | |||
AL833713.1 | - | 3 | 269 | |||
BC131725.1 | - | 12 | 1395 | 40123868 | ||
CR749459.1 | - | 7 | 743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
