siRNA Details
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 19 | 4021 | ||
NM_207325.2 | NP_997208.2 | 19 | 4047 | |||
XM_006723045.1 | XP_006723108.1 | 16 | 3563 | |||
XM_011526526.2 | XP_011524828.1 | 19 | 4002 | |||
XM_017026366.1 | XP_016881855.1 | 18 | 3936 | |||
XM_017026367.1 | XP_016881856.1 | 19 | 3961 | |||
GenBank mRNA | AK125846.1 | - | 3 | 2165 | ||
AK129497.1 | - | 19 | 4002 | |||
AL833713.1 | - | 9 | 4000 | |||
CR749459.1 | - | 13 | 3217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
