siRNA Details
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 19 | 2608 | ||
NM_207325.2 | NP_997208.2 | 19 | 2634 | |||
XM_006723045.1 | XP_006723108.1 | 16 | 2150 | |||
XM_011526526.2 | XP_011524828.1 | 19 | 2589 | |||
XM_017026366.1 | XP_016881855.1 | 18 | 2523 | |||
XM_017026367.1 | XP_016881856.1 | 19 | 2548 | |||
GenBank mRNA | AK125846.1 | - | 3 | 752 | ||
AK126264.1 | - | 15 | 2762 | |||
AK126757.1 | - | 19 | 2625 | |||
AK129497.1 | - | 19 | 2589 | |||
AL833713.1 | - | 9 | 2587 | |||
BC131725.1 | - | 18 | 2456 | 40123868 | ||
CR749459.1 | - | 13 | 1804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
