siRNA Details
Gene Symbol: JPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020433.4 | NP_065166.2 | 1 | 1009 | ||
NM_175913.3 | NP_787109.2 | 1 | 1009 | |||
XM_006723833.3 | XP_006723896.1 | 1 | 1001 | |||
GenBank mRNA | AL132999.1 | - | 1 | 524 | ||
BC029957.1 | - | 1 | 1009 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
