siRNA Details
Gene Symbol: TMCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008740.3 | NP_001008740.1 | 2 | 526 | ||
GenBank mRNA | BC108666.1 | - | 2 | 459 | 5743890 | |
BU567634.1 | - | 2 | 529 | |||
BU569147.1 | - | 2 | 495 | |||
DB518323.1 | - | 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
