siRNA Details
Gene Symbol: LAMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005559.3 | NP_005550.2 | 55 | 7944 | ||
XM_011525655.2 | XP_011523957.1 | 55 | 7938 | |||
XM_011525656.2 | XP_011523958.1 | 45 | 6359 | |||
GenBank mRNA | AF351616.1 | - | 2 | 1428 | ||
AK091949.1 | - | 6 | 1257 | |||
BC039051.1 | - | 7 | 868 | |||
BC062318.1 | - | 3 | 304 | |||
BC144590.1 | - | 54 | 8865 | |||
X58531.1 | - | 55 | 7944 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
