siRNA Details
Gene Symbol: HMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005519.1 | NP_005510.1 | 1 | 273 | ||
XM_005269743.3 | XP_005269800.1 | 2 | 135 | |||
GenBank mRNA | BC132758.1 | - | 1 | 83 | 40146780 | |
BC137139.1 | - | 1 | 83 | 9021136 | ||
BQ929484.1 | - | 1 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
