siRNA Details
Gene Symbol: VWA5B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138345.2 | NP_612354.1 | 1, 2 | 246 | ||
XM_011513288.2 | XP_011511590.1 | 2, 3 | 870 | |||
XM_011513289.2 | XP_011511591.1 | 2, 3 | 953 | |||
XM_011513290.2 | XP_011511592.1 | 2, 3 | 416 | |||
XM_011513291.2 | XP_011511593.1 | 1, 2 | 1082 | |||
XM_011513292.2 | XP_011511594.1 | 1, 2 | 1082 | |||
GenBank mRNA | BC136609.1 | - | 1, 2 | 246 | 9020599 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
