siRNA Details
Gene Symbol: SCAND2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003654.2 | 1 | 517 | |||
NR_004859.1 | 1 | 517 | ||||
GenBank mRNA | AF229246.1 | - | 1 | 532 | ||
AK022844.1 | - | 1 | 517 | |||
AK290489.1 | - | 1 | 527 | |||
BC011547.1 | - | 1 | 499 | |||
BC012929.1 | - | 1 | 499 | |||
BC063836.1 | - | 1 | 473 | |||
BC093922.1 | - | 1 | 150 | |||
BC093924.1 | - | 1 | 150 | |||
BC143241.1 | - | 1 | 150 | |||
BC143243.1 | - | 1 | 150 | 9051744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
