siRNA Details
Gene Symbol: CIDEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279.3 | NP_001270.1 | 5 | 812 | ||
NM_001318383.1 | NP_001305312.1 | 5 | 1305 | |||
NR_134607.1 | 6 | 1572 | ||||
GenBank mRNA | AK122762.1 | - | 6 | 1588 | ||
BC031896.1 | - | 5 | 877 | 4776542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
