siRNA Details
Gene Symbol: CIDEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279.3 | NP_001270.1 | 2 | 134 | ||
NM_001318383.1 | NP_001305312.1 | 2 | 627 | |||
NR_134607.1 | 2 | 627 | ||||
GenBank mRNA | AF041378.1 | - | 2 | 69 | ||
AK122762.1 | - | 2 | 643 | |||
AY364639.1 | - | 2 | 69 | |||
BC031896.1 | - | 2 | 199 | 4776542 | ||
BQ718688.1 | - | 2 | 82 | |||
R73455.1 | - | 2 | 134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
