siRNA Details
Gene Symbol: ST7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017744.4 | NP_060214.2 | 4 | 776 | ||
NM_138727.3 | NP_620055.1 | 4 | 725 | |||
NM_138728.2 | NP_620056.1 | 4 | 776 | |||
NM_138729.3 | NP_620057.1 | 4 | 776 | |||
XM_005270963.4 | XP_005271020.1 | 4 | 299 | |||
XM_005270964.3 | XP_005271021.1 | 3 | 391 | |||
XM_011541627.2 | XP_011539929.1 | 4 | 485 | |||
XM_011541628.2 | XP_011539930.1 | 4 | 485 | |||
XM_011541629.2 | XP_011539931.1 | 4 | 499 | |||
XM_011541630.2 | XP_011539932.1 | 4 | 1195 | |||
XM_011541631.2 | XP_011539933.1 | 4 | 323 | |||
XM_017001534.1 | XP_016857023.1 | 4 | 485 | |||
XM_017001535.1 | XP_016857024.1 | 4 | 485 | |||
XM_017001536.1 | XP_016857025.1 | 3 | 391 | |||
XM_017001538.1 | XP_016857027.1 | 3 | 266 | |||
GenBank mRNA | AB081317.1 | - | 4 | 547 | ||
AB081318.1 | - | 4 | 496 | |||
AB081319.1 | - | 4 | 547 | |||
AB081320.1 | - | 4 | 547 | |||
AK021719.1 | - | 3 | 386 | |||
AK095837.1 | - | 2 | 84 | |||
AK291042.1 | - | 4 | 477 | |||
AK295955.1 | - | 4 | 474 | |||
AK301869.1 | - | 4 | 477 | |||
AK304004.1 | - | 4 | 434 | |||
AK308727.1 | - | 3 | 400 | |||
BC027898.1 | - | 3 | 378 | |||
BC037830.1 | - | 4 | 1189 | 4812133 | ||
BC047392.1 | - | 4 | 502 | 5273549 | ||
BC065501.1 | - | 4 | 683 | 4417786 | ||
BX537988.1 | - | 5 | 638 | |||
BX640667.1 | - | 4 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
