siRNA Details
Gene Symbol: PRELP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002725.3 | NP_002716.1 | 2 | 862 | ||
NM_201348.1 | NP_958505.1 | 2 | 852 | |||
GenBank mRNA | BC032498.1 | - | 2 | 780 | 5222817 | |
CR541787.1 | - | 1 | 662 | |||
CR542270.1 | - | 1 | 662 | |||
U29089.1 | - | 2 | 790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
