siRNA Details
Gene Symbol: LOC728613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003713.1 | 1 | 86 | |||
GenBank mRNA | AK307076.1 | - | 1 | 65 | ||
AK308990.1 | - | 1 | 28 | |||
AK311210.1 | - | 1 | 84 | |||
AK311251.1 | - | 1 | 90 | |||
AK315872.1 | - | 1 | 50 | |||
BC014604.1 | - | 1 | 67 | |||
BC053586.1 | - | 1 | 16 | |||
BC067863.1 | - | 1 | 56 | |||
DA383480.1 | - | 1 | 86 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
