siRNA Details
Gene Symbol: PRDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007257.2 | NP_001007258.1 | 3 | 1958 | ||
NM_012231.4 | NP_036363.2 | 8 | 3218 | |||
NM_015866.4 | NP_056950.2 | 8 | 3218 | |||
XM_017002253.1 | XP_016857742.1 | 7 | 2440 | |||
XM_017002254.1 | XP_016857743.1 | 7 | 2440 | |||
XM_017002255.1 | XP_016857744.1 | 7 | 2450 | |||
XM_017002256.1 | XP_016857745.1 | 8 | 2519 | |||
XM_017002257.1 | XP_016857746.1 | 7 | 2448 | |||
XM_017002258.1 | XP_016857747.1 | 5 | 2150 | |||
XM_017002259.1 | XP_016857748.1 | 4 | 1979 | |||
XM_017002260.1 | XP_016857749.1 | 3 | 2345 | |||
XM_017002261.1 | XP_016857750.1 | 3 | 2060 | |||
XM_017002262.1 | XP_016857751.1 | 4 | 1979 | |||
XM_017002263.1 | XP_016857752.1 | 3 | 1873 | |||
GenBank mRNA | AB208877.1 | - | 3 | 1937 | ||
AK293725.1 | - | 4 | 1794 | |||
AK300441.1 | - | 4 | 1666 | |||
AK308135.1 | - | 4 | 1791 | |||
BX647310.1 | - | 4 | 1981 | |||
D45132.1 | - | 3 | 1939 | |||
U17838.1 | - | 8 | 3220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
