siRNA Details
Gene Symbol: COL17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000494.3 | NP_000485.3 | 11 | 952 | ||
GenBank mRNA | AB900157.1 | - | 10 | 783 | ||
AK309940.1 | - | 11 | 898 | |||
BC004478.2 | - | 11 | 934 | |||
M91669.1 | - | 11 | 888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
