siRNA Details
Gene Symbol: TRBV5-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028083.1 | - | 3, 4 | 584 |
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF430686.1 | - | 2, 3 | 279 |
Gene Symbol: IL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ506993.1 | - | 2, 3 | 344 |
Gene Symbol: TRBV21-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF088323.1 | - | 108 | |||
AF088332.1 | - | 119 | ||||
AF088359.1 | - | 120 | ||||
AF088369.1 | - | 118 | ||||
AF088441.1 | - | 2, 3 | 120 |
Gene Symbol: TRBV23-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF088485.1 | - | 2, 3 | 134 | ||
AF088491.1 | - | 2, 3 | 127 | |||
AF088500.1 | - | 2, 3 | 122 | |||
AF088521.1 | - | 108 | ||||
AF088532.1 | - | 113 | ||||
AF088539.1 | - | 129 | ||||
AF088547.1 | - | 2, 3 | 85 | |||
AF088548.1 | - | 2, 3 | 140 | |||
AF088551.1 | - | 2, 3 | 124 |
Gene Symbol: TRBV3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC070387.1 | - | 3, 4 | 468 |
Gene Symbol: TRBV6-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC110303.1 | - | 3, 4 | 461 |
Gene Symbol: TRBV7-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC108321.1 | - | 3, 4 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
