siRNA Details
Gene Symbol: CAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206747.1 | NP_001193676.1 | 3 | 1131 | ||
NM_001206748.1 | NP_001193677.1 | 2 | 1016 | |||
NM_001233.4 | NP_001224.1 | 3 | 1131 | |||
NM_198212.2 | NP_937855.1 | 2 | 943 | |||
GenBank mRNA | AF035752.1 | - | 3 | 970 | ||
AK310786.1 | - | 2 | 804 | |||
AV650449.1 | - | 1 | 498 | |||
AY353255.1 | - | 3 | 962 | |||
BC005256.1 | - | 3 | 1131 | 3829012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
