siRNA Details
Gene Symbol: CAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF179102.1 | - | 7 | 772 |
Gene Symbol: CAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206747.1 | NP_001193676.1 | 3 | 626 | ||
NM_001206748.1 | NP_001193677.1 | 2 | 511 | |||
NM_001233.4 | NP_001224.1 | 3 | 626 | |||
NM_198212.2 | NP_937855.1 | 2 | 438 | |||
GenBank mRNA | AB451285.1 | - | 3 | 445 | ||
AB451417.1 | - | 3 | 445 | |||
AF035752.1 | - | 3 | 465 | |||
AK310786.1 | - | 2 | 299 | |||
AK311964.1 | - | 3 | 468 | |||
AY353255.1 | - | 3 | 457 | |||
BC005256.1 | - | 3 | 626 | 3829012 | ||
BT007051.1 | - | 3 | 445 | |||
CR407685.1 | - | 3 | 445 | |||
DA882274.1 | - | 2 | 511 |
Gene Symbol: LOC105375463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF070114.1 | - | 11 | 1147 | ||
EF070118.1 | - | 5 | 547 | |||
EF070121.1 | - | 7 | 666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
