siRNA Details
Gene Symbol: RBM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015014.2 | NP_055829.2 | 11 | 1341 | ||
NR_027762.1 | 10 | 1204 | ||||
XM_011544133.1 | XP_011542435.1 | 10 | 1641 | |||
XM_017000721.1 | XP_016856210.1 | 11 | 1698 | |||
GenBank mRNA | AK292362.1 | - | 11 | 1327 | ||
BC029451.1 | - | 11 | 1341 | 4809263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
