siRNA Details
Gene Symbol: CDHR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007540.3 | NP_001007541.2 | 3 | 309 | ||
XM_011533700.2 | XP_011532002.1 | 4 | 438 | |||
XM_011533701.2 | XP_011532003.1 | 4 | 370 | |||
XM_017006366.1 | XP_016861855.1 | 5 | 668 | |||
XM_017006367.1 | XP_016861856.1 | 5 | 632 | |||
XM_017006368.1 | XP_016861857.1 | 5 | 668 | |||
XM_017006369.1 | XP_016861858.1 | 5 | 653 | |||
XM_017006370.1 | XP_016861859.1 | 5 | 668 | |||
XM_017006371.1 | XP_016861860.1 | 5 | 668 | |||
GenBank mRNA | AY358221.1 | - | 3 | 309 | ||
BC132756.1 | - | 3 | 309 | |||
BC144445.1 | - | 3 | 309 | 9052970 | ||
BC146674.1 | - | 3 | 295 | 40148497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
