siRNA Details
Gene Symbol: CDHR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358221.1 | - | 3 | 731 | ||
BC132756.1 | - | 3 | 731 | |||
BC144445.1 | - | 4 | 655 | 9052970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
