siRNA Details
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178134.1 | NP_001171605.1 | 4 | 650 | ||
NM_199168.3 | NP_954637.1 | 3 | 1381 | |||
GenBank mRNA | BC031072.1 | - | 3 | 1373 | ||
BC039893.1 | - | 3 | 1359 | 5729604 | ||
DQ345517.1 | - | 4 | 602 | |||
L36034.1 | - | 3 | 1366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
