siRNA Details
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199168.3 | NP_954637.1 | 3 | 363 | ||
GenBank mRNA | AK292628.1 | - | 3 | 366 | ||
AY874118.1 | - | 3 | 350 | |||
BC031072.1 | - | 3 | 355 | |||
BC039893.1 | - | 3 | 341 | 5729604 | ||
CD172464.1 | - | 3 | 361 | |||
DA266752.1 | - | 3 | 364 | |||
L36034.1 | - | 3 | 350 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021704, AK045092, BC020297, BC029335, D21072, and others.
Gene Ontology Categories:
