siRNA Details
Gene Symbol: MBNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306070.1 | NP_001292999.1 | 8 | 4441 | ||
NM_144778.3 | NP_659002.1 | 9 | 4529 | |||
NM_207304.2 | NP_997187.1 | 7 | 4398 | |||
XM_005254018.2 | XP_005254075.1 | 10 | 4868 | |||
XM_006719912.2 | XP_006719975.1 | 9 | 4773 | |||
XM_011521045.2 | XP_011519347.1 | 9 | 4380 | |||
XM_017020308.1 | XP_016875797.1 | 10 | 4475 | |||
XM_017020309.1 | XP_016875798.1 | 8 | 4310 | |||
GenBank mRNA | AF339782.1 | - | 1 | 1468 | ||
AF491866.1 | - | 9 | 4479 | |||
CR749802.1 | - | 8 | 4357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
