siRNA Details
Gene Symbol: HMCES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006109.1 | NP_001006109.1 | 7 | 1451 | ||
NM_020187.2 | NP_064572.2 | 7 | 1278 | |||
XM_005247636.3 | XP_005247693.1 | 7 | 1463 | |||
XM_005247637.3 | XP_005247694.1 | 6 | 1187 | |||
XM_017006877.1 | XP_016862366.1 | 7 | 1295 | |||
GenBank mRNA | AF035296.1 | - | 1 | 263 | ||
AF201934.1 | - | 7 | 1292 | |||
AK025883.1 | - | 6 | 1123 | |||
AK225264.1 | - | 6 | 1147 | |||
AK297501.1 | - | 6 | 1150 | |||
AY007144.1 | - | 3 | 729 | |||
BC009993.2 | - | 7 | 1358 | 4130099 | ||
BC010125.2 | - | 7 | 1241 | 3945258 | ||
BC050686.1 | - | 7 | 1451 | 6155077 | ||
BC088363.1 | - | 7 | 1275 | 30398247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
