siRNA Details
Gene Symbol: NKAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007531.2 | NP_001007532.1 | 1 | 977 | ||
GenBank mRNA | BC038240.1 | - | 1 | 907 | 5766894 | |
BC101681.1 | - | 1 | 935 | 8069187 | ||
BC101685.1 | - | 1 | 935 | 8069191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
