siRNA Details
Gene Symbol: LBH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030915.3 | NP_112177.2 | 3 | 2376 | ||
XM_017005059.1 | XP_016860548.1 | 3 | 2534 | |||
GenBank mRNA | AF110224.1 | - | 1 | 1146 | ||
AF258555.1 | - | 1 | 1367 | |||
AK023425.1 | - | 1 | 1443 | |||
AK027232.1 | - | 3 | 2168 | |||
AL136703.1 | - | 3 | 2381 | |||
BC109376.1 | - | 3 | 2380 | 6495944 | ||
CN485291.1 | - | 1 | 241 | |||
EF025586.1 | - | 3 | 2353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
