siRNA Details
Gene Symbol: ZNF93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031218.3 | NP_112495.2 | 4 | 1156 | ||
GenBank mRNA | AK022550.1 | - | 1 | 63 | ||
AK096342.1 | - | 4 | 1113 | |||
AK295921.1 | - | 1 | 488 | |||
BC039001.1 | - | 1 | 132 | |||
BC136718.1 | - | 4 | 1008 | 9020708 | ||
BC136719.1 | - | 4 | 1008 | 9020709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
