siRNA Details
Gene Symbol: ZNF93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031218.3 | NP_112495.2 | 4 | 1931 | ||
GenBank mRNA | AK096342.1 | - | 5 | 1804 | ||
AK295921.1 | - | 1 | 1263 | |||
BC136718.1 | - | 4 | 1783 | 9020708 | ||
BC136719.1 | - | 4 | 1783 | 9020709 | ||
BU165449.1 | - | 1 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
