siRNA Details
Gene Symbol: OR6C74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005490.1 | NP_001005490.1 | 1 | 127 | ||
XM_017019126.1 | XP_016874615.1 | 2 | 208 | |||
GenBank mRNA | BC140754.1 | - | 1 | 217 | 9021622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
