siRNA Details
Gene Symbol: CORO2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003389.3 | NP_003380.3 | 10 | 1358 | ||
NM_052820.3 | NP_438171.1 | 10 | 1208 | |||
XM_011518986.2 | XP_011517288.1 | 10 | 1535 | |||
GenBank mRNA | AK292788.1 | - | 10 | 1152 | ||
BC000010.1 | - | 9 | 1096 | 3504229 | ||
BC011690.2 | - | 10 | 1174 | 3350035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
