siRNA Details
Gene Symbol: FGD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018351.3 | NP_060821.3 | 9 | 3347 | ||
GenBank mRNA | AB037783.1 | - | 8 | 1120 | ||
AK002045.1 | - | 2 | 138 | |||
AK124904.1 | - | 9 | 3347 | |||
AK128492.1 | - | 8 | 2224 | |||
BC013319.1 | - | 3 | 230 | |||
BC125225.1 | - | 8 | 750 | |||
BC125226.1 | - | 8 | 750 | |||
BX538322.1 | - | 9 | 3263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
