siRNA Details
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243266.1 | NP_001230195.1 | 6 | 764 | ||
NM_024021.3 | NP_076926.2 | 8 | 1069 | |||
NM_148975.2 | NP_683876.1 | 7 | 923 | |||
XM_017017909.1 | XP_016873398.1 | 7 | 936 | |||
GenBank mRNA | AB013102.1 | - | 7 | 764 | ||
AF068288.1 | - | 7 | 857 | |||
AF237912.1 | - | 7 | 874 | |||
AF350500.1 | - | 6 | 827 | |||
AK075081.1 | - | 8 | 954 | |||
AK292089.1 | - | 7 | 878 | |||
BC020648.1 | - | 7 | 798 | 4720088 | ||
CB046393.1 | - | 6 | 633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
