siRNA Details
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243266.1 | NP_001230195.1 | 6 | 985 | ||
NM_024021.3 | NP_076926.2 | 8 | 1290 | |||
NM_148975.2 | NP_683876.1 | 7 | 1144 | |||
XM_017017909.1 | XP_016873398.1 | 7 | 1157 | |||
GenBank mRNA | AF237912.1 | - | 7 | 1095 | ||
AK075081.1 | - | 8 | 1175 | |||
AK292089.1 | - | 7 | 1099 | |||
BC020648.1 | - | 7 | 1019 | 4720088 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
