siRNA Details
Gene Symbol: SBK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199824.2 | NP_001186753.1 | 4 | 453 | ||
XM_011526298.2 | XP_011524600.1 | 4 | 544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
