siRNA Details
Gene Symbol: SBK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101401.2 | NP_001094871.2 | 1 | 233 | ||
XM_006723327.3 | XP_006723390.1 | 2 | 342 | |||
XM_011527227.2 | XP_011525529.1 | 2 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
