siRNA Details
Gene Symbol: DPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135155.2 | NP_001128627.1 | 12 | 1608 | ||
NM_001135156.2 | NP_001128628.1 | 12 | 1622 | |||
NM_001289978.1 | NP_001276907.1 | 12 | 1638 | |||
NM_004647.3 | NP_004638.2 | 11 | 1506 | |||
XM_005259289.2 | XP_005259346.1 | 11 | 1557 | |||
XM_005259290.3 | XP_005259347.1 | 12 | 1579 | |||
XM_005259292.4 | XP_005259349.1 | 11 | 1494 | |||
XM_006723407.2 | XP_006723470.1 | 12 | 1609 | |||
XM_006723408.3 | XP_006723471.1 | 12 | 1577 | |||
XM_006723409.2 | XP_006723472.1 | 12 | 1624 | |||
XM_006723410.2 | XP_006723473.1 | 9 | 1176 | |||
XM_011527356.1 | XP_011525658.1 | 11 | 1587 | |||
XM_011527357.2 | XP_011525659.1 | 11 | 1527 | |||
XM_011527358.2 | XP_011525660.1 | 12 | 1653 | |||
GenBank mRNA | AK094632.1 | - | 12 | 1935 | ||
AK127198.1 | - | 8 | 2288 | |||
BC021191.2 | - | 12 | 1582 | |||
U43843.1 | - | 11 | 1401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
