siRNA Details
Gene Symbol: B3GNT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033309.2 | NP_171608.2 | 2 | 2107 | ||
GenBank mRNA | AF171091.1 | - | 1 | 689 | ||
AY358485.1 | - | 2 | 2039 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
