siRNA Details
Gene Symbol: COLEC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130386.2 | NP_569057.1 | 10 | 2457 | ||
XM_011525741.2 | XP_011524043.1 | 9 | 2315 | |||
GenBank mRNA | AB005145.1 | - | 10 | 2312 | ||
AB038518.1 | - | 10 | 2450 | |||
AB052103.1 | - | 8 | 3840 | |||
AL713657.1 | - | 6 | 1269 | |||
BC060789.1 | - | 10 | 2429 | 30344546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
