siRNA Details
Gene Symbol: COLEC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130386.2 | NP_569057.1 | 5 | 1329 | ||
XM_011525741.2 | XP_011524043.1 | 4 | 1187 | |||
GenBank mRNA | AB005145.1 | - | 5 | 1184 | ||
AB034251.1 | - | 4 | 1114 | |||
AB038518.1 | - | 5 | 1322 | |||
AB052103.1 | - | 4 | 1116 | |||
AL713657.1 | - | 1 | 141 | |||
BC060789.1 | - | 5 | 1301 | 30344546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
