siRNA Details
Gene Symbol: OR7D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175883.2 | NP_787079.1 | 1 | 662 | ||
GenBank mRNA | AK095468.1 | - | 1 | 662 | ||
X64975.1 | - | 1 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
