siRNA Details
Gene Symbol: C15orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032413.3 | NP_115789.1 | 3 | 432 | ||
NM_197955.2 | NP_922946.1 | 4 | 318 | |||
GenBank mRNA | AB026707.1 | - | 4 | 212 | ||
AF228422.1 | - | 3 | 353 | |||
AK026298.1 | - | 3 | 350 | |||
BC021173.2 | - | 4 | 238 | 5269504 | ||
BM312626.1 | - | 4 | 282 | |||
CA865860.1 | - | 4 | 324 | |||
DB447060.1 | - | 4 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
