siRNA Details
Gene Symbol: C15orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032413.3 | NP_115789.1 | 2, 3 | 386 | ||
NM_197955.2 | NP_922946.1 | 3, 4 | 272 | |||
GenBank mRNA | AB026707.1 | - | 3, 4 | 166 | ||
AF228422.1 | - | 2, 3 | 307 | |||
AK026298.1 | - | 2, 3 | 304 | |||
BC021173.2 | - | 3, 4 | 192 | 5269504 | ||
BM312626.1 | - | 3, 4 | 236 | |||
CA865860.1 | - | 3, 4 | 278 | |||
DB447060.1 | - | 3, 4 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
