siRNA Details
Gene Symbol: RBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323281.1 | NP_001310210.1 | 8 | 1590 | ||
NM_001323282.1 | NP_001310211.1 | 9 | 1655 | |||
NM_002895.4 | NP_002886.2 | 9 | 1517 | |||
NM_183404.3 | NP_899662.1 | 9 | 1517 | |||
XM_006723842.2 | XP_006723905.1 | 8 | 1228 | |||
XM_011528955.2 | XP_011527257.1 | 9 | 1293 | |||
XM_011528958.2 | XP_011527260.1 | 9 | 1293 | |||
XM_017027992.1 | XP_016883481.1 | 10 | 1473 | |||
GenBank mRNA | AK290380.1 | - | 9 | 1293 | ||
AK308096.1 | - | 9 | 1278 | |||
AK310870.1 | - | 9 | 1440 | |||
BC032247.1 | - | 9 | 1300 | 4521266 | ||
L14812.1 | - | 9 | 1298 | |||
M74547.1 | - | 7 | 831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
