siRNA Details
Gene Symbol: RBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323281.1 | NP_001310210.1 | 11 | 1907 | ||
NM_001323282.1 | NP_001310211.1 | 12 | 1972 | |||
NM_002895.4 | NP_002886.2 | 12 | 1834 | |||
NM_183404.3 | NP_899662.1 | 12 | 1834 | |||
XM_006723842.2 | XP_006723905.1 | 11 | 1545 | |||
XM_011528955.2 | XP_011527257.1 | 12 | 1610 | |||
XM_011528958.2 | XP_011527260.1 | 12 | 1610 | |||
XM_017027992.1 | XP_016883481.1 | 13 | 1790 | |||
GenBank mRNA | AK290380.1 | - | 12 | 1610 | ||
AK308096.1 | - | 12 | 1595 | |||
BC032247.1 | - | 12 | 1617 | 4521266 | ||
L14812.1 | - | 12 | 1615 | |||
M74547.1 | - | 10 | 1148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
