siRNA Details
Gene Symbol: SHOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000451.3 | NP_000442.1 | 2 | 941 | ||
NM_006883.2 | NP_006874.1 | 2 | 941 | |||
GenBank mRNA | U89331.1 | - | 2 | 943 | ||
Y11535.1 | - | 1 | 341 | |||
Y11536.1 | - | 1 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
