siRNA Details
Gene Symbol: PYHIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152501.4 | NP_689714.2 | 3 | 568 | ||
NM_198928.4 | NP_945146.1 | 3 | 541 | |||
NM_198929.4 | NP_945147.1 | 3 | 568 | |||
NM_198930.3 | NP_945148.1 | 3 | 541 | |||
XM_005244930.1 | XP_005244987.1 | 3 | 568 | |||
XM_011509242.2 | XP_011507544.1 | 3 | 568 | |||
XM_011509243.2 | XP_011507545.1 | 3 | 568 | |||
XM_017000463.1 | XP_016855952.1 | 3 | 568 | |||
GenBank mRNA | AK024890.1 | - | 1 | 27 | ||
AK308350.1 | - | 3 | 547 | |||
AY185344.1 | - | 3 | 555 | |||
AY185345.1 | - | 3 | 528 | |||
AY185346.1 | - | 3 | 555 | |||
AY185347.1 | - | 3 | 528 | |||
BC020822.1 | - | 3 | 547 | |||
BC090944.1 | - | 3 | 550 | 5212268 | ||
BC139741.1 | - | 3 | 495 | 40033401 | ||
DB126431.1 | - | 3 | 564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
